• USA Home
  • Search Results

  • dmt2otbdmsrctacphosphoramidite10503014998966211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-2′O-TBDMS-rC(tac) Phosphoramidite

5 Products
Synonym:   DMT-2′-O-TBDMS-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-?methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]
Image of DMT-2′O-TBDMS-rC(tac) Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C57H76N5O10PSi
Molecular Weight:
 1050.30
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More