• USA Home
  • Search Results

  • dmt2otcruphosphoramidite92403121908992511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-2′O-TC-rU Phosphoramidite

4 Products
Synonym:   O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)uridine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)
Image of DMT-2′O-TC-rU Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C44H54N5O11PS2
Molecular Weight:
 924.03
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More