• USA Home
  • Search Results

  • dmtbiotinc6phosphoramidite8460713513787011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-Biotin-C6 Phosphoramidite

3 Products
Synonym:   5′ Biotin Amidite, 5′-Biotin Phosphoramidite, 6-[(2-Cyanoethoxy)diisopropylaminophosphanoxy]-[1-N-(4,4′-dimethoxytrityl)biotinyl]aminohexane, DMT-Biotin-C6 Amidite, N-DMT-Biotinyl-2-Aminoethoxyethanol
Image of DMT-Biotin-C6 Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C46H64N5O6PS
Molecular Weight:
 846.07
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Compatibility
  Product Line
Show More