• USA Home
  • Search Results

  • dmtdcbzphosphoramidite8339110221298611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-dC(bz) Phosphoramidite

8 Products
Synonym:   Cytidine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-dC(bz) amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-?bis(1-methylethyl)phosphoramidite], N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]
Image of DMT-dC(bz) Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C46H52N5O8P
Molecular Weight:
 833.91
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Nucleoside
  Physical Form
  UV-Absorption Condition
  Type
  Titertest
  Product Line
  Color
  Biological Source
  Compatibility
  Package quantity
Show More