• USA Home
  • Search Results

  • dmtdtcpgcolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-dT-CPG column

18 Products
Image of DMT-dT-CPG column
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Compatibility
  UV-Absorption Condition
  Type
  Package quantity
  Capacity
  Assay
  Product Line
  Pore size
Show More

Product #
Description
T421010 DMT-dT-CPG column, pore size 1000 Å, 0.2 μmol, configured for PerkinElmer 8900,
T3110MM-HH DMT-dT-CPG column, pore size 500 Å, 50 nmol, configured for MerMade,
T3210MM-HH DMT-dT-CPG column, pore size 500 Å, 0.2 μmol, configured for MerMade,
T341010 DMT-dT-CPG column, pore size 500 Å, 50 nmol, configured for ABI, configured for PerkinElmer,
T361010 DMT-dT-CPG column, pore size 500 Å, 1 μmol, configured for ABI, configured for PerkinElmer,
T4110MM-HH DMT-dT-CPG column, pore size 1000 Å, 50 nmol, configured for MerMade,
T4210MM-HH DMT-dT-CPG column, pore size 1000 Å, 0.2 μmol, configured for MerMade,
T311080 DMT-dT-CPG column, pore size 500 Å, 50 nmol, configured for PerkinElmer,
T321080 DMT-dT-CPG column, pore size 500 Å, 0.2 μmol, configured for PerkinElmer,
T331080 DMT-dT-CPG column, pore size 500 Å, 1 μmol, configured for PerkinElmer,
T421080 DMT-dT-CPG column, pore size 1000 Å, 0.2 μmol, configured for PerkinElmer,
T431080 DMT-dT-CPG column, pore size 1000 Å, 1 μmol, configured for PerkinElmer,
T311040 DMT-dT-CPG column, pore size 500 Å, 50 nmol, configured for MerMade,
T311060 DMT-dT-CPG column, pore size 500 Å, 50 nmol, configured for PerkinElmer, configured for ABI,
T321060 DMT-dT-CPG column, pore size 500 Å, 0.2 μmol, configured for PerkinElmer, configured for ABI,
T331060 DMT-dT-CPG column, pore size 500 Å, 1 μmol, configured for PerkinElmer, configured for ABI,
T421060 DMT-dT-CPG column, pore size 1000 Å, 0.2 μmol, configured for PerkinElmer, configured for ABI,
T431060 DMT-dT-CPG column, pore size 1000 Å, 1 μmol, configured for PerkinElmer, configured for ABI,