• USA Home
  • Search Results

  • dmtlocabzphosphoramidite8859420605579011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-locA(bz) Phosphoramidite

3 Products
CAS Number:
Empirical Formula (Hill Notation):
 C48H52N7O8P
Molecular Weight:
 885.94
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Compatibility
  Absorption
  Physical Form
  Solubility
  Product Line
  Color
  Biological Source
Show More