• USA Home
  • Search Results

  • dnorleucine1311732756011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Norleucine

1 Products
Synonym:   (R)-2-Aminocaproic acid, (R)-2-Aminohexanoic acid
Image of <SC>D</SC>-Norleucine
CAS Number:
Empirical Formula (Hill Notation):
 C6H13NO2
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 1721749
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More

Product #
Description
N6627 D-Norleucine,