• USA Home
  • Search Results

  • dnorvaline11715201312911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Norvaline

1 Products
Synonym:   (R)-(−)-2-Aminopentanoic acid
Image of <SC>D</SC>-Norvaline
CAS Number:
Linear Formula:
 CH3CH2CH2CH(NH2)CO2H
Molecular Weight:
 117.15
 Beilstein Registry Number:
 1721161
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
851620 D-Norvaline, 99%,