• USA Home
  • Search Results

  • docosane3106062997011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Docosane

4 Products
Image of Docosane
CAS Number:
Linear Formula:
 CH3(CH2)20CH3
Molecular Weight:
 310.60
 EC Index Number
 211-121-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More