• USA Home
  • Search Results

  • dodecanethiolfunctionalizedgoldnanoparticles1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dodecanethiol functionalized gold nanoparticles

2 Products
Synonym:   Dodecylthiol functionalized gold nanoparticles
Image of Dodecanethiol functionalized gold nanoparticles
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Functional group
  Physical Form
  Particle Dimension
  ID clarifier
Show More