• USA Home
  • Search Results

  • dodin28744243910311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dodin

2 Products
Synonym:   1-Dodecyl-guanidiniumacetat
Image of Dodin
CAS Number:
Empirical Formula (Hill Notation):
 C15H33N3O2
Molecular Weight:
 287.44
 Beilstein/REAXYS Number:
 6546960
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More