• USA Home
  • Search Results

  • doramectin8991111770425311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Doramectin

1 Products
Synonym:   25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin
Image of Doramectin
CAS Number:
Empirical Formula (Hill Notation):
 C50H74O14
Molecular Weight:
 899.11
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More