• USA Home
  • Search Results

  • doxycyclinehyclate512942439014511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Doxycycline hyclate

5 Products
Synonym:   Doxycycline hydrochloride hemiethanolate hemihydrate
Image of Doxycycline hyclate
CAS Number:
Empirical Formula (Hill Notation):
 C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
Molecular Weight:
 512.94
 Beilstein Registry Number:
 5702728
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Color
  Biological Source
  Assay
  Mode of Action
  Solubility
  Anal. Standard Form
  Activity Spectrum
  Special Grade
Show More