• USA Home
  • Search Results

  • doxylaminesuccinatesalt3884656210711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Doxylamine succinate salt

1 Products
Image of Doxylamine succinate salt
CAS Number:
Empirical Formula (Hill Notation):
 C17H22N2O · C4H6O4
Molecular Weight:
 388.46
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D3775 Doxylamine succinate salt,