• USA Home
  • Search Results

  • dpenicillamine149215267511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Penicillamine

1 Products
Synonym:   3,3-Dimethyl-D-cysteine, 3-Mercapto-D-valine, D-(−)-2-Amino-3-mercapto-3-methylbutanoic acid
Image of <SC>D</SC>-Penicillamine
CAS Number:
Linear Formula:
 (CH3)2C(SH)CH(NH2)CO2H
Molecular Weight:
 149.21
 Beilstein/REAXYS Number:
 1722375
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P4875 D-Penicillamine, 98-101%,