• USA Home
  • Search Results

  • dpinitol194181028463611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Pinitol

2 Products
Synonym:   3-O-Methyl-D-chiro-inositol
Image of <SC>D</SC>-Pinitol
CAS Number:
Empirical Formula (Hill Notation):
 C7H14O6
Molecular Weight:
 194.18
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More