• USA Home
  • Search Results

  • drierite13614777818911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Drierite

6 Products
Synonym:   Calcium sulfate, Gypsum
Image of Drierite<SUP>™</SUP>
CAS Number:
Linear Formula:
 CaSO4
Molecular Weight:
 136.14
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Application(s)
  Physical Form
Show More