• USA Home
  • Search Results

  • dttp504151842343311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

dTTP

1 Products
Synonym:   2′-deoxy-thymidine-5′-triphosphate, TTP, Thymidine 5′-triphosphate sodium salt, dTTP, deoxy-thymidine-5′-triphosphate, 2′-
CAS Number:
Empirical Formula (Hill Notation):
 C10H16N2O14P3Na
Molecular Weight:
 504.15
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
11051482001 dTTP, Lithium salt,