• USA Home
  • Search Results

  • duroquinone1642052717311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Duroquinone

2 Products
Synonym:   2,3,5,6-Tetramethyl-1,4-benzoquinone, Tetramethyl-p-benzoquinone
Image of Duroquinone
CAS Number:
Empirical Formula (Hill Notation):
 C10H12O2
Molecular Weight:
 164.20
 Beilstein/REAXYS Number:
 1909128
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More