• USA Home
  • Search Results

  • dutasteride5285316465623911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dutasteride

3 Products
Synonym:   (5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost-1-ene-17-carboxamide, 17β-N-[2,5-Bis(trifluoromethyl)phenyl]carbamoyl-4-aza-5α-androst-1-en-3-one
Image of Dutasteride
CAS Number:
Empirical Formula (Hill Notation):
 C27H30F6N2O2
Molecular Weight:
 528.53
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More