• USA Home
  • Search Results

  • dutp46814freeacidbasis9473609111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

dUTP

1 Products
Synonym:   2′-Deoxyuridine 5′-triphosphate sodium salt solution, dUTP sodium salt
Image of dUTP
CAS Number:
Empirical Formula (Hill Notation):
 C9H15N2O14P3 · xNa+
Molecular Weight:
 468.14 (free acid basis)
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More