• USA Home
  • Search Results

  • dysprosium16250742991611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dysprosium

31 Products
Image of Dysprosium
CAS Number:
Empirical Formula (Hill Notation):
 Dy
Molecular Weight:
 162.50
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Reaction Suitability
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Assay
Show More

Product #
Description
261076 Dysprosium, ingot, 99.9% trace rare earth metals basis,
263028 Dysprosium, chips, 99.9% trace rare earth metals basis,
GF87262364 Dysprosium, foil, thickness 0.2 mm, size 25 × 25 mm, purity 99%,
GF74526880 Dysprosium, foil, thickness 0.2 mm, size 25 × 75 mm, purity 99%,
GF91494108 Dysprosium, lump, 25 mm max. lump size, weight 100 g, purity 99%,
GF07796212 Dysprosium, foil, thickness 0.2 mm, size 25 × 50 mm, purity 99%,
GF56950937 Dysprosium, foil, thickness 0.50 mm, size 25 × 25 mm, tolerance 0.2,
GF57559337 Dysprosium, foil, thickness 0.50 mm, size 50 × 50 mm, tolerance 0.2,
GF11898096 Dysprosium, foil, 100x100mm, thickness 0.125mm, as rolled, 99%,
GF69984769 Dysprosium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%,
GF89687222 Dysprosium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%,
GF85958229 Dysprosium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%,
GF48095185 Dysprosium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%,
GF10903676 Dysprosium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%,
GF65566536 Dysprosium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%,
GF69010602 Dysprosium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%,
GF04383592 Dysprosium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%,
GF13501358 Dysprosium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%,
GF89822065 Dysprosium, foil, not light tested, 5x5mm, thickness 0.005mm, as rolled, 99%,
GF56066171 Dysprosium, powder, 10g, max. particle size 500 micron, 99.9%,
GF54698451 Dysprosium, powder, 1g, max. particle size 500 micron, 99.9%,
GF87801723 Dysprosium, powder, 5g, max. particle size 500 micron, 99.9%,
GF74506899 Dysprosium, rod, 100mm, diameter 6.35mm, cast, 99%,
GF40435039 Dysprosium, rod, 50mm, diameter 2.0mm, cast, 99%,
GF30196054 Dysprosium, rod, 50mm, diameter 6.35mm, cast, 99%,
GF56425451 Dysprosium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%,
GF15989978 Dysprosium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%,
GF99502836 Dysprosium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%,
GF22672380 Dysprosium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%,
GF00748515 Dysprosium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%,
GF34048526 Dysprosium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%,