• USA Home
  • Search Results

  • dysprosiumiiinitratehydrate34851anhydrousbasis10064113211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dysprosium(III) nitrate hydrate

1 Products
Image of Dysprosium(III) nitrate hydrate
CAS Number:
Linear Formula:
 Dy(NO3)3·xH2O
Molecular Weight:
 348.51 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More