• USA Home
  • Search Results

  • e2bromo2butene13500301771811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(E)-2-Bromo-2-butene

1 Products
Image of (<I>E</I>)-2-Bromo-2-butene
CAS Number:
Linear Formula:
 CH3CH=C(Br)CH3
Molecular Weight:
 135.00
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
657484 (E)-2-Bromo-2-butene, 96%,