• USA Home
  • Search Results

  • ecaprolactam1131610560211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ε-Caprolactam

4 Products
Synonym:   2-Oxohexamethyleneimine, Aza-2-cycloheptanone, epsilon-Caprolactam
Image of ε-Caprolactam
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO
Molecular Weight:
 113.16
 Beilstein/REAXYS Number:
 106934
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More