• USA Home
  • Search Results

  • echinacoside786738285437311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Echinacoside

4 Products
Image of Echinacoside
CAS Number:
Empirical Formula (Hill Notation):
 C35H46O20
Molecular Weight:
 786.73
 Beilstein Registry Number:
 4778612
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
Show More