• USA Home
  • Search Results

  • eicosane2825511295811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eicosane

3 Products
Image of Eicosane
CAS Number:
Linear Formula:
 CH3(CH2)18CH3
Molecular Weight:
 282.55
 Beilstein/REAXYS Number:
 1700722
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
  Application(s)
Show More