• USA Home
  • Search Results

  • element14pdms1000123456314862911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Element14® PDMS 1000

1 Products
Synonym:   Dimethylpolysiloxane, Poly(dimethylsiloxane)
Image of Element14<SUP>®</SUP> PDMS 1000
CAS Number:
Linear Formula:
 [Si(CH3)2O]n
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Viscosity
Show More