• USA Home
  • Search Results

  • eleutherosideb3723711834311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eleutheroside B

3 Products
Synonym:   (E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-β-D-glucopyranoside, Ilexanthin A, Ligustrin, Lilacin, Magnolenin, Methoxyconiferine, Sinapyl alcohol 4-O-glucoside, Syringin, Syringoside
Image of Eleutheroside B
CAS Number:
Empirical Formula (Hill Notation):
 C17H24O9
Molecular Weight:
 372.37
 Beilstein/REAXYS Number:
 97166
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Assay
Show More