• USA Home
  • Search Results

  • embelin2943955024311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Embelin

2 Products
Synonym:   2,5-Dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, Embelic acid, Emberine
Image of Embelin
CAS Number:
Empirical Formula (Hill Notation):
 C17H26O4
Molecular Weight:
 294.39
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Solubility
  Assay
Show More