• USA Home
  • Search Results

  • enalaprilmaleate492527609516411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Enalapril maleate

4 Products
Synonym:   (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt, Enalapril maleate salt
Image of Enalapril maleate
CAS Number:
Empirical Formula (Hill Notation):
 C20H28N2O5 · C4H4O4
Molecular Weight:
 492.52
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Anal. Standard Form
  Physical Form
  Manufacturer designation
Show More