• USA Home
  • Search Results

  • endothelin3humanrat26430411739993611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Endothelin 3 human, rat

1 Products
Synonym:   ET-3
Image of Endothelin 3 human, rat
CAS Number:
Empirical Formula (Hill Notation):
 C121H168N26O33S4
Molecular Weight:
 2643.04
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More