• USA Home
  • Search Results

  • eosiny647891508694911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eosin Y

2 Products
Synonym:   2′,4′,5′,7′-Tetrabromofluorescein, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, Eosin yellowish, Solvent red 43
Image of Eosin Y
CAS Number:
Empirical Formula (Hill Notation):
 C20H8Br4O5
Molecular Weight:
 647.89
 Colour Index Number:
 45380:2
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Solubility
Show More