• USA Home
  • Search Results

  • ephedrinehydrochloride2016913471411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ephedrine hydrochloride

1 Products
Image of Ephedrine hydrochloride
CAS Number:
Linear Formula:
 C6H5CH(OH)CH(CH3)NHCH3 · HCl
Molecular Weight:
 201.69
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More