• USA Home
  • Search Results

  • epichlorohydrin925210689811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Epichlorohydrin

2 Products
Synonym:   (±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane
Image of (±)-Epichlorohydrin
CAS Number:
Empirical Formula (Hill Notation):
 C3H5ClO
Molecular Weight:
 92.52
 Beilstein/REAXYS Number:
 79785
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Purity Grade
  Refractive Index
  Assay
Show More