• USA Home
  • Search Results

  • epidermalgrowthfactorfrommurinesubmaxillarygland123456222950911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Epidermal Growth Factor from murine submaxillary gland

2 Products
Synonym:   EGF, Epidermal cell growth factor
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.77
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Physical Form
  Application(s)
  Potency
  Package quantity
  Color
Show More