• USA Home
  • Search Results

  • epigallocatechin3062797074111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-Epigallocatechin

4 Products
Synonym:   (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane
Image of (−)-Epigallocatechin
CAS Number:
Empirical Formula (Hill Notation):
 C15H14O7
Molecular Weight:
 306.27
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More