• USA Home
  • Search Results

  • epinephrinehydrochloride2196732963511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Epinephrine hydrochloride

1 Products
Synonym:   (±)-Adrenalin, 4-(1-Hydroxy-2-[methylamino]ethyl)-1,2-benzenediol hydrochloride
Image of (±)-Epinephrine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C9H13NO3 · HCl
Molecular Weight:
 219.67
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More