• USA Home
  • Search Results

  • ergocalciferol396655014611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ergocalciferol

5 Products
Synonym:   Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2
Image of Ergocalciferol
CAS Number:
Empirical Formula (Hill Notation):
 C28H44O
Molecular Weight:
 396.65
 Beilstein/REAXYS Number:
 1916682
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
 eCl@ss
 34058017
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Anal. Standard Form
  Assay
  Physical Form
  Special Grade
  Manufacturer designation
  Color
Show More