• USA Home
  • Search Results

  • ergocalciferolforsystemsuitability396655014611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ergocalciferol for system suitability

1 Products
Synonym:   Calciferol, Ercalciol, Ergocalciferol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2
Image of Ergocalciferol for system suitability
CAS Number:
Empirical Formula (Hill Notation):
 C28H44O
Molecular Weight:
 396.65
 Beilstein/REAXYS Number:
 1916682
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More