• USA Home
  • Search Results

  • ergosterol396655787411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ergosterol

4 Products
Synonym:   3β-Hydroxy-5,7,22-ergostatriene, 5,7,22-Ergostatrien-3β-ol, Provitamin D2
Image of Ergosterol
CAS Number:
Empirical Formula (Hill Notation):
 C28H44O
Molecular Weight:
 396.65
 Beilstein/REAXYS Number:
 2338604
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Functional group
  Manufacturer designation
  Assay
  Anal. Standard Form
  Color
  Biological Source
  Application(s)
  Physical Form
Show More