• USA Home
  • Search Results

  • eriocitrin596531346328011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eriocitrin

4 Products
Synonym:   (S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[6-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictioside, Eriodictyol 7-O-rutinoside
Image of Eriocitrin
CAS Number:
Empirical Formula (Hill Notation):
 C27H32O15
Molecular Weight:
 596.53
 Beilstein/REAXYS Number:
 1304401
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
  Application(s)
  Manufacturer designation
Show More