• USA Home
  • Search Results

  • eriodictyol28825404938111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Eriodictyol

3 Products
Synonym:   3′,4′,5,7-Tetrahydroxyflavanone
Image of (±)-Eriodictyol
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O6
Molecular Weight:
 288.25
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More