• USA Home
  • Search Results

  • eriodictyol2882555258911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eriodictyol

2 Products
Synonym:   (S)-3′,4′,5,7-Tetrahydroxyflavanone, Eriodictiol, Huazhongilexone
Image of Eriodictyol
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O6
Molecular Weight:
 288.25
 Beilstein/REAXYS Number:
 5104930
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More