• USA Home
  • Search Results

  • erucicacid3385711286711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Erucic acid

3 Products
Synonym:   cis-13-Docosenoic acid, Prifac 2990
Image of Erucic acid
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)11COOH
Molecular Weight:
 338.57
 Beilstein/REAXYS Number:
 1728049
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Biological Source
  Functional group
  Physical Form
Show More