• USA Home
  • Search Results

  • erythromycinethylsuccinate86205126462611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Erythromycin ethyl succinate

5 Products
Image of Erythromycin ethyl succinate
CAS Number:
Empirical Formula (Hill Notation):
 C43H75NO16
Molecular Weight:
 862.05
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Anal. Standard Form
Show More