• USA Home
  • Search Results

  • esmololhydrochloride331838116117311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Esmolol hydrochloride

2 Products
Synonym:   4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzenepropanoic acid methyl ester hydrochloride hydrochloride
Image of Esmolol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C16H26NClO4
Molecular Weight:
 331.83
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More