• USA Home
  • Search Results

  • estriol288385027111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Estriol

6 Products
Synonym:   1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene
Image of Estriol
CAS Number:
Empirical Formula (Hill Notation):
 C18H24O3
Molecular Weight:
 288.38
 Beilstein/REAXYS Number:
 2508172
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
  Manufacturer designation
Show More