• USA Home
  • Search Results

  • estrone270375316711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Estrone

6 Products
Synonym:   1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin
Image of Estrone
CAS Number:
Empirical Formula (Hill Notation):
 C18H22O2
Molecular Weight:
 270.37
 Beilstein/REAXYS Number:
 1915077
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Agency/Method
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More