• USA Home
  • Search Results

  • estropipate43656728037711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Estropipate

3 Products
Synonym:   3-(Sulfooxy)-estra-1,3,5(10)-trien-17-one : piperazine (1:1), Estrone sulfate piperazine salt, Harmogen, Ogen
Image of Estropipate
CAS Number:
Empirical Formula (Hill Notation):
 C18H22O5S·C4H10N2
Molecular Weight:
 436.56
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More